Learn More
Specifications
Specifications
| Assay Range | 15.60 to 1000 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 10216 |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | bG174L6.2, bG174L6.2 (MSF: megakaryocyte stimulating factor), CACP, FLJ32635, HAPO, Hemangiopoietin, JCAP, lubricin, megakaryocyte stimulating factor, Megakaryocyte-stimulating factor, MSF, MSFproteoglycan 4, (megakaryocyte stimulating factor, articular superficial zoneprotein, camptodactyly, arthropathy, coxa vara, pericarditis syndrome), PRG4, Proteoglycan 4, Superficial zone proteoglycan, SZP, SZPHAPOarticular superficial zone protein |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
