Learn More
Clontech Labs 3P HUMAN LUNG RNA 50UG
SDP
Supplier:  Clontech Labs 3P 636524
Encompass
Human Lung Total RNA, 50 microgram, These RNA can be used for a variety of applications, including Atlas Array hybridizations, SMART PCR cDNA synthesis, and RT-PCR analysis
Catalog No. NC9946720
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.