Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human LXR beta Synthetic Peptide

Catalog No. PEP1000
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1000 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1000 Supplier Invitrogen™ Supplier No. PEP1000
Add to Cart
Add to Cart

Synthetic Peptide

LXR-B Synthetic Peptide, PEP-1000, from Invitrogen.

The liver X receptors, LXRA and LXRB, form a subfamily of the nuclear receptor superfamily and are key regulators of macrophage function, controlling transcriptional programs involved in lipid homeostasis and inflammation. The inducible LXRA is highly expressed in liver, adrenal gland, intestine, adipose tissue, macrophages, lung, and kidney, whereas LXRB is ubiquitously expressed. Ligand-activated LXRs form obligate heterodimers with retinoid X receptors and regulate expression of target genes containing LXR response elements.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 7376
Species Human
Purification Method purified
Accession Number P55055
Common Name Human LXR beta
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol NR1H2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.