Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Human Lymphatic System Anatomical Model
SCI_ED

Catalog No. S012563 Shop All Eisco Products
Change view
Click to view available options
Dimensions (L x W x H): 18 x 8 x 2 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S012563 Dimensions (L x W x H): 18 x 8 x 2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S012563 Supplier Eisco™ Supplier No. AM0348
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The lymphatic system is an integral part of the immune system and it plays an important part in the body to defense against diseases. Includes a key card identifying 9 labeled structures.

  • LYMPHATIC SYSTEM - Human figure mounted on base depicting structures of the human lymphatic system, include major ducts, glands, and vessels
  • EXPERTLY DESIGNED - Designed by medical professionals and educators for detail and accuracy
  • QUALITY CONSTRUCTION - Custom molded using the highest quality nontoxic polymers and polyvinyl chloride (PVC) for durability
  • HAND-PAINTED - Hand-painted by skilled artists to deliver the highest level of detail possible using vibrant paint
  • NUMBERED STRUCTURES - Nine important structures are numbered on the model and easily identified with the included color key card

Specifications

Color Tan
Dimensions (L x W x H) 18 x 8 x 2 in.
Material PVC
Weight (English) 1.8 lb
Gender Male, Female
Product Type Lymphatic System Model
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.