Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Lymphotactin (XCL1) Recombinant Protein, PeproTech®

Catalog No. 300205UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
20 μg
250 μg
5 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-20-5UG 5 μg
30020100UG 100 μg
300-20-20UG 20 μg
30020250UG 250 μg
30020500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 300-20-5UG Supplier Gibco™ Supplier No. 300205UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

300-20-1MG will be provided as 2 x 500 μg (300-20-500UG). Recombinant Human Lymphotactin is a 10.0 kDa protein consisting of 92 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Lymphotactin, otherwise known as XCL1, is a member of the C-chemokine family that uniquely acts as a chemoattractant for lymphocytes, but not monocytes or neutrophils, suggesting an important regulatory role in lymphocyte trafficking and inflammation. Lymphotactin, shown to signal through chemokine XC receptor 1 (XCR1), is detected at the highest level in the spleen and at low levels in the lungs, small intestine and colon, and is abundantly and selectively expressed by activated T cells, in particular CD8+ T cells, and is also produced by NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P47992
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6375
Molecular Weight (g/mol) 10 kDa
Name Human Lymphotactin (XCL1)
Quantity 5 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI661682; ATAC; C motif chemokine 1; chemokine (C motif) ligand 1; cytokine SCM-1; Lptn; LTN; Lymphotactin; lymphotaxin; SCM1; SCM-1; SCM1A; SCM-1 A; SCM1alpha; SCM-1 alpha; SCM-1-alpha; SCM1beta; SCM-1 beta; SCM-1-beta; SCYC1; single cysteine motif 1 A; small inducible cytokine subfamily C, member 1 (lymphotactin); small-inducible cytokine C1; XC chemokine ligand 1; X-C motif chemokine ligand 1; XCL1
Common Name Lymphotactin
Gene Symbol Xcl1
Biological Activity Determined by its ability to bind alpha9 integrin expressed on the surface of MOLT4 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence GSEVSDKRTC VSLTTQRLPV SRIKTYTITE GSLRAVIFIT KRGLKVCADP QATWVRDVVR SMDRKSNTRN NMIQTKPTGT QQSTNTAVTL TG
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.