Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Lysosomal alpha-Glucosidase Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7182524
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8329SP 25 μg
MAB8329100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8329SP Supplier R&D Systems Supplier No. MAB8329SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

LYAG/GAA Monoclonal specifically detects LYAG/GAA in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen LYAG/GAA
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 2489C
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. P10253
Gene Alias Acid alpha-Glucosidase, Acid maltase, Aglucosidase alfa, EC 3.2.1.20, GAA, glucosidase, alpha; acid, LYAG, lysosomal alpha-glucosidase, Lysosomal alphaGlucosidase
Host Species Rabbit
Immunogen Human embryonic kidney cell, HEK293-derived human Lysosomal alpha-Glucosidase protein, Ala70-Cys952, Accession # P10253
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2548
Test Specificity Detects human Lysosomal alpha-Glucosidase protein in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.