Learn More
American Research Products Inc Human Lysozyme LZM ELISA Kit
Supplier:  American Research Products Inc CSBE09135H
Encompass
Human Lysozyme LZM ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates urine Method Sandwich
Catalog No. 50-003-29473