Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Lysyl Oxidase Homolog 2 DuoSet ELISA
SDP

Catalog No. DY263905 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY263905 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY263905 Supplier R&D Systems™ Supplier No. DY263905

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)

Human Lysyl Oxidase Homolog 2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Lysyl Oxidase Homolog 2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Lysyl Oxidase Homolog 2/LOXL2.

Specifications

Assay Range 0.375 to 24 ng/mL
Detectable Analytes Lysyl Oxidase Homolog 2
Assay Solid Phase ELISA
Conjugate Biotin
Cross Reactivity <50%cross-species reactivity observed with species tested
Immunoassay Kit Format 96-well Strip Plate
Product Line R&D Systems
Product Type ELISA Kit
Sample Type Cell Culture Supernatant
Sufficient For To run ELISAs on at least 5 x 96-well plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 4017
Host Species Human
Quantity 5 Plates
Sample Volume 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.