Learn More
ACCEGEN BIOTECH Human Mammary Epithelial Cells, ≥0.5 million/ frozen vial
Supplier:  ACCEGEN BIOTECH ABCTC3691
Encompass
Human mammary epithelial cells (HMECs) sit within duct and lobes of breast and play an important role in maintaining breast function and facilitate milk production during lactation. Mammary epithelium consists of an inner luminal epithelial cell layer and an outer myoepithelial cell layer adjacent to basement membrane. The luminal lineage can be divided into ductal and alveolar cells while the contractile myoepithelial cells are smooth muscle-like cells found at the basal surface of epithelium. Female breast epithelial cells undergo reorganization during puberty, pregnancy, lactation, and menopause in response to various hormones and cytokine signals, making it susceptible to development of breast cancer.Why Choose Human Mammary Epithelial Cells, Adult from AcceGenAcceGen Human mammary epithelial cells are isolated from adult female breast tissue. Cells are cryopreserved at passage 5 and delivered on dry ice.
Catalog No. NC2749584
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.