Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Human Mandible & Lower Jaw Model
SCI_ED

Catalog No. S27428 Shop All Eisco Products
Change view
Click to view available options
Dimensions (L x W x H):
4.25 x 4.5 x 3.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S27428 4.25 x 4.5 x 3.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27428 Supplier Eisco™ Supplier No. AMCH1016AS
Add to Cart
Add to Cart

Provides an incredible amount of realism and detail including texture and bony landmarks, includes 16 teeth

  • Anatomically correct-model
  • Natural size and color
  • Realistic details, textures, and bony landmarks
  • Ideal for educational and medical demonstrations
  • Made of durable, washable PVC plastic to withstand years of student and instructor use
  • Use for individual study or as replacement for missing/damaged pieces to a full skeletal model set

Specifications

Color Natural
Dimensions (L x W x H) 4.25 x 4.5 x 3.25 in.
For Use With (Application) Educational and Medical Demonstrations
Includes 16 Teeth
Material PVC
Product Type Anatomical Model, Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.