Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human MAP2 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NB013322 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013322 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013322 Supplier Novus Biologicals™ Supplier No. NBP280251
Add to Cart
Add to Cart

This kit recognizes Human MAP-2 in samples.

No significant cross-reactivity or interference between Human MAP-2 and analogues was observed.

Specifications

Accession Number P11137, P11137
Assay Range 62.5 to 4000 pg/mL
Assay Sensitivity 37.5 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 4133
Gene Symbol MAP2
Host Species Human
Quantity 1 Kit
Research Discipline Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
Target MAP2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.