Learn More
Abnova™ Human MAP3K7 (NP_663304.1, 1 a.a. - 303 a.a.) and TAB1 (NP_006107.1, 437 a.a. - 504 a.a.) Full-length Recombinant Protein with His tag expressed in baculovirus infected Sf21 cells
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene was identified as a regulator of the MAP kinase kinase kinase MAP3K7/TAK1, which is known to mediate various intracellular signaling pathways, such as those induced by TGF beta, interleukin 1, and WNT-1. This protein interacts and thus activates TAK1 kinase. It has been shown that the C-terminal portion of this protein is sufficient for binding and activation of TAK1, while a portion of the N-terminus acts as a dominant-negative inhibitor of TGF beta, suggesting that this protein may function as a mediator between TGF beta receptors and TAK1. This protein can also interact with and activate the mitogen-activated protein kinase 14 (MAPK14/p38alpha), and thus represents an alternative activation pathway, in addition to the MAPKK pathways, which contributes to the biological responses of MAPK14 to various stimuli. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq]
Specifications
Specifications
| Accession Number | NP_663304.1 (Gene ID : 6885);NP_006107.1 (Gene ID : 10454) |
| For Use With (Application) | Functional Study, SDS-PAGE |
| Formulation | Liquid |
| Gene ID (Entrez) | 6885/10454 |
| Molecular Weight (g/mol) | 45kDa |
| Name | MAP3K7/TAB1 (Human) Recombinant Protein |
| Preparation Method | Baculovirus infected insect cell (Sf21) expression system |
| Purification Method | Ni-NTA affinity chromatography |
| Quality Control Testing | Loading 1 ug protein in SDS-PAGE |
| Quantity | 5 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.