Learn More
Sino Biological Human MAPK14 MAb 20UL 110435-R0004-20
Supplier:  Sino Biological 110435R000420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Thr180, Tyr182 of human Phospho-p38 MAPK.
Catalog No. 50-241-2054
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.