Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MAPK8IP2 Full-length ORF (AAH47527.1, 1 a.a. - 824 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89951776 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-951-776 10 μg
89-951-777 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-951-776 Supplier Abnova™ Supplier No. H00023542P02S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is closely related to MAPK8IP1/IB1/JIP-1, a scaffold protein that is involved in the c-Jun amino-terminal kinase signaling pathway. This protein is expressed in brain and pancreatic cells. It has been shown to interact with, and regulate the activity of MAPK8/JNK1, and MAP2K7/MKK7 kinases. This protein thus is thought to function as a regulator of signal transduction by protein kinase cascade in brain and pancreatic beta-cells. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene. [provided by RefSeq]

Sequence: MADRAEMFSLSTFHSLSPPGCRPPQDISLEEFDDEDLSEITDDCGLGLSYDSDHCEKDSLSLGRSEQPHPICSFQDDFQGFEMIDDNEEEDDEDEEEEEEEEEGDGEGQEGGDPGSEAPAPGPLIPSASVEEPHKHRPTTLRLTTLGAQDSLNNNGGFDLVRPASWQETALCSPAPEALRELPGPLPATDTGPGGAQSPVRPGCDCEGNRPAEPPAPGGTSPSSDPGIEADLRSRSSGGRGGRRSSQELSSPGSDSEDAGGARLGRMISSISETELELSSDGGSSSSGRSSHLTNSIEEASSPASEPEPPREPPRRPAFLPVGPDDTNSEYESGSESEPDLSEDADSPWLLSNLVSRMISEGSSPIRCPGQCLSPAPRPPGEPVSPAGGAAQDSQDPEAAAGPGGVELVDMETLCAPPPPAPAAPRPGPAQPGPCLFLSNPTRDTITPLWAAPGRAARPGRACSAACSKEEDEEDDEEEEDAEDSAGSPGGRGTGPSAPRDASLVYDAVKYTLVVDEHTQLELVSLRRCAGLGHDSEEDSGGEASEEEAGAALLGGGQVSGDTSPDSPDLTFSKKFLNVFVNSTSRSSSTESFGLFSCLVNGEEREQTHRAVFRFIPRHPDELELDVDDPVLVEAEEDDFWFRGFNMRTGERGVFPAFYAHAVPGPAKDLLGSKRSPCWVERFDVQFLGSVEVPCHQGNGILCAAMQKIATARKLTVHLRPPASCDLEISLRGVKLSLSGGGPEFQRCSHFFQMKNISFCGCHPRNSCYFGFITKHPLLSRFACHVFVSQESMRPVAQSVGRAFLEYYQEHLAYACPTEDIYLE

Specifications

Accession Number AAH47527.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23542
Molecular Weight (g/mol) 114.3kDa
Name MAPK8IP2 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MADRAEMFSLSTFHSLSPPGCRPPQDISLEEFDDEDLSEITDDCGLGLSYDSDHCEKDSLSLGRSEQPHPICSFQDDFQGFEMIDDNEEEDDEDEEEEEEEEEGDGEGQEGGDPGSEAPAPGPLIPSASVEEPHKHRPTTLRLTTLGAQDSLNNNGGFDLVRPASWQETALCSPAPEALRELPGPLPATDTGPGGAQSPVRPGCDCEGNRPAEPPAPGGTSPSSDPGIEADLRSRSSGGRGGRRSSQELSSPGSDSEDAGGARLGRMISSISETELELSSDGGSSSSGRSSHLTNSIEEASSPASEPEPPREPPRRPAFLPVGPDDTNSEYESGSESEPDLSEDADSPWLLSNLVSRMISEGSSPIRCPGQCLSPAPRPPGEPVSPAGGAAQDSQDPEAAAGPGGVELVDMETLCAPPPPAPAAPRPGPAQPGPCLFLSNPTRDTITPLWAAPGRAARPGRACSAACSKEEDEEDDEEEEDAEDSAGSPGGRGTGPSAPRDASLVYDAVKYTLVVDEHTQLELVSLRRCAGLGHDSEEDSGGEASEEEAGAALLGGGQVSGDTSPDSPDLTFSKKFLNVFVNSTSRSSSTESFGLFSCLVNGEEREQTHRAVFRFIPRHPDELELDVDDPVLVEAEEDDFWFRGFNMRTGERGVFPAFYAHAVPGPAKDLLGSKRSPCWVERFDVQFLGSVEVPCHQGNGILCAAMQKIATARKLTVHLRPPASCDLEISLRGVKLSLSGGGPEFQRCSHFFQMKNISFCGCHPRNSCYFGFITKHPLLSRFACHVFVSQESMRPVAQSVGRAFLEYYQEHLAYACPTEDIYLE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias IB2/JIP2/PRKM8IPL
Common Name MAPK8IP2
Gene Symbol MAPK8IP2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.