Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MARK2 (NP_059672.2, 1 a.a. - 745 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells

Catalog No. 89951861 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-951-861 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-951-861 Supplier Abnova™ Supplier No. P5594
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

This gene encodes a member of the Par-1 family of serine/threonine protein kinases. The protein is an important regulator of cell polarity in epithelial and neuronal cells, and also controls the stability of microtubules through phosphorylation and inactivation of several microtubule-associating proteins. The protein localizes to cell membranes. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Accession Number NP_059672.2
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 2011
Molecular Weight (g/mol) 110kDa
Name MARK2 (Human) Recombinant Protein
Preparation Method Baculovirus infected insect cell (Sf21) expression system
Purification Method Glutathione sepharose chromatography
Quality Control Testing Loading 1 ug protein in SDS-PAGE
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias EMK1/MGC99619/PAR-1/Par1b
Common Name MARK2
Gene Symbol MARK2
Biological Activity The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: CHKtide. ATP: 100 uM.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Baculovirus infected insect cell (Sf21) expression system
Form Liquid
Purity or Quality Grade 63% by SDS-PAGE/CBB staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.