Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MBL Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7061028
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2307SP 25 μg
AF2307 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2307SP Supplier R&D Systems Supplier No. AF2307SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 6 publications

MBL Polyclonal specifically detects MBL in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen MBL
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH96182
Gene Alias COLEC1, COLEC1collectin-1, Collectin-1, HSMBPC, Mannan-binding protein, Mannose-binding lectin, mannose-binding lectin (protein C) 2, soluble, mannose-binding lectin (protein C) 2, soluble (opsonic defect), mannose-binding protein C, MBL2, MBL2D, MBLmannan-binding lectin, MBP, MBP1, MBP1mannose-binding lectin 2, soluble (opsonic defect), MBP-C, MGC116832, MGC116833
Gene Symbols MBL2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human MBL Glu21-Ile248 Accession # AAH96182
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4153
Test Specificity Detects human MBL in ELISAs and Western blots. In sandwich ELISAs, less than 0.05% cross-reactivity with recombinant mouse (rm) MBL and rmMBL-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.