Learn More
Specifications
Specifications
| Assay Range | 93.80 - 6000 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| Gene ID (Entrez) | 4155 |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | Hmbpr, MGC99675, mLd, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein, shi |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
