Learn More
Description
- Also known as: HDMX; hdm2; ACTFS
- MDM2 is a nuclear-localized E3 ubiquitin ligase.
- It can promote tumor formation by targeting tumor suppressor proteins, such as p53, for proteasomal degradation.
- Overexpression or amplification of this locus is detected in a variety of different cancers.
- Alternatively spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | MDM2/HDM2 |
| Accession Number | AAM78554 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1489 bp |
| Gene ID (Entrez) | 4193 |
| Gene Alias | ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
