Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MEF2A Full-length ORF (AAH13437, 1 a.a. - 499 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89952136 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-952-136 25 ug
89-952-135 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-952-136 Supplier Abnova™ Supplier No. H00004205P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The process of differentiation from mesodermal precursor cells to myoblasts has led to the discovery of a variety of tissue-specific factors that regulate muscle gene expression. The myogenic basic helix-loop-helix proteins, including myoD (MIM 159970), myogenin (MIM 159980), MYF5 (MIM 159990), and MRF4 (MIM 159991) are one class of identified factors. A second family of DNA binding regulatory proteins is the myocyte-specific enhancer factor-2 (MEF2) family. Each of these proteins binds to the MEF2 target DNA sequence present in the regulatory regions of many, if not all, muscle-specific genes. The MEF2 genes are members of the MADS gene family (named for the yeast mating type-specific transcription factor MCM1, the plant homeotic genes 'agamous' and 'deficiens' and the human serum response factor SRF (MIM 600589)), a family that also includes several homeotic genes and other transcription factors, all of which share a conserved DNA-binding domain.[supplied by OMIM]

Sequence: MGRKKIQITRIMDERNRQVTFTKRKFGLMKKAYELSVLCDCEIALIIFNSSNKLFQYASTDMDKVLLKYTEYNEPHESRTNSDIVEALNKKEHRGCDSPDPDTSYVLTPHTEEKYKKINEEFDNMMRNHKIAPGLPPQNFSMSVTVPVTSPNALSYTNPGSSLVSPSLAASSTLTDSSMLSPPQTTLHRNVSPGAPQRPPSTGNAGGMLSTTDLTVPNGAGSSPVGNGFVNSRASPNLIGATGANSLGKVMPTKSPPPPGGGNLGMNSRRPDLRVVIPPSSKGMMPPLNTQRISSSQATQPLATPVVSVTTPSLPPQGLVYSAMPTAYNTDYSLTSADLSALQGFNSPGMLSLGQVSAWQQHHLGQAALSSLVAGGQLSQGSNLSINTNQNISIKSEPISPPRDRMTPSGFQQQQQQQQQQQPPPPPQPQPQPPQPQPRQEMGRSPVDSLSSSSSSYDGSDREDPRGDFHSPIVLGRPPNTEDRESPSVKRMRMDAWVT

Specifications

Accession Number AAH13437
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4205
Molecular Weight (g/mol) 80.41kDa
Name MEF2A (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MGRKKIQITRIMDERNRQVTFTKRKFGLMKKAYELSVLCDCEIALIIFNSSNKLFQYASTDMDKVLLKYTEYNEPHESRTNSDIVEALNKKEHRGCDSPDPDTSYVLTPHTEEKYKKINEEFDNMMRNHKIAPGLPPQNFSMSVTVPVTSPNALSYTNPGSSLVSPSLAASSTLTDSSMLSPPQTTLHRNVSPGAPQRPPSTGNAGGMLSTTDLTVPNGAGSSPVGNGFVNSRASPNLIGATGANSLGKVMPTKSPPPPGGGNLGMNSRRPDLRVVIPPSSKGMMPPLNTQRISSSQATQPLATPVVSVTTPSLPPQGLVYSAMPTAYNTDYSLTSADLSALQGFNSPGMLSLGQVSAWQQHHLGQAALSSLVAGGQLSQGSNLSINTNQNISIKSEPISPPRDRMTPSGFQQQQQQQQQQQPPPPPQPQPQPPQPQPRQEMGRSPVDSLSSSSSSYDGSDREDPRGDFHSPIVLGRPPNTEDRESPSVKRMRMDAWVT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ADCAD1/RSRFC4/RSRFC9
Common Name MEF2A
Gene Symbol MEF2A
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.