Learn More
Sino Biological Anti-Meprin beta Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10708RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MEP1B / Meprin beta (rh MEP1B / Meprin beta; Catalog#10708-H08H; NP_005916.2; Met1-Thr652). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9696