Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Mesenchymal Stem Cell Functional Identification Kit
SDP

Catalog No. SC006 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SC006 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SC006 Supplier R&D Systems™ Supplier No. SC006

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Adipogenic Supplement, Osteogenic Supplement, Chondrogenic Supplement, ITS Supplement, Goat anti-mouse FABP-4, Mouse anti-human Osteocalcin, Goat anti-human Aggrecan

To verify multipotency of human mesenchymal stem cells by in vitro functional differentiation

Can be used with the Human MSC Verification Flow Kit (Mfr. No. FMC020) to define MSCs according to the ISCT recommendations
Reliably induces MSC trilineage differentiation using defined supplements
Includes premium quality antibodies to confirm differen

Specifications

Cross Reactivity Human
Quantity 1 Kit
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Concentrate
Product Line R&D Systems
Product Type Stem Cell Differentiation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.