Learn More
Human Mesenchymal Stem Cell Lineage Antibody Cocktail w/Isotype Control, BD
Shop All BD Biosciences ProductsDescription
Host Species:
Clone: hMSC Negative Cocktail: 581, ICRF44, HIB19, HI30, G46-6
hMSC Isotype Control Negative Cocktail: X40, G155-178
Cross Reactivity Species: Hu
Flow Cytometry
Specifications
Specifications
| Conjugate | PE |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.