Learn More
R&D Systems™ Human Mesenchymal Stem Cell Marker Antibody Panel
SDP
Catalog No. SC017 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SC017 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. SC017 Supplier R&D Systems™ Supplier No. SC017
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
25μg each of antibodies to Stro-1, CD90, CD106, CD105, CD146, CD166, CD44, CD19, and CD45
For the verification of mesenchymal stem/stromal cell identity by flow cytometry
Researchers use different techniques to isolate, culture, and differentiate human mesenchymal stem/stromal cells (MSCs). Variations in experimental approaches as well as differences in the MSC starting population may account for experimental variability a
Cost-effective panel of premium quality antibodies for the positive and negative identification of MSCs
Uses 9 markers to increase confidence in MSC identification
Enables verification of MSC identity in less than 2 hours with use of fluorochrome-conjug
Specifications
Quantity 1 Kit
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems