Learn More
R&D Systems™ Human Mesenchymal Stem Cell Verification Flow Kit
SDP
Catalog No. FMC020 Shop All Novus Products
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Catalog No. Content And Storage
FMC020 Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 options
This item is not returnable. View return policy
Catalog No. FMC020 Supplier R&D Systems™ Supplier No. FMC020
Add to Cart
Add to Cart
This item is not returnable. View return policy
Provides single-step staining for the verification of human MSCs
Mesenchymal stromal cells are a heterogeneous population of cultured cells that are adherent to plastic, have a distinct morphology, and express a specific set of marker proteins. Within this heterogeneous population are mesenchymal stem cells. Mesenchymal stem cells are multipotent, self-renewing cells that have the ability to differentiate into adipocytes, chondrocytes, and osteoblasts when cultured in vitro.
Antibodies for MSC verification according to the markers proposed by the International Society for Cellular Therapy
Includes all of the antibodies needed to fulfill the ISCT minimal criteria for the definition of human MSCs
Can be used with the Human MSC Functional ID kit to define MSCs according to the ISCT recommendations
Provides a cost-effective method to analyze several MSC markers in a single sample
Reveals heterogeneity in the starting cell population
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems