Learn More
ACROBiosystems Human Mesothelin 10mg
Supplier:  ACROBiosystems MBSC00310MG
Encompass_Preferred
ActiveMax Human MSLN uBeads premium grade (for cells) are uniform superparamagnetic beads of 5.5 µm with streptavidin on its surface and coupled with biotinylated Human MSLN Protein expressed from human 293 cells (HEK293) and contains AA Glu 296 - Gly 580 (Accession # AAH09272.1).
Catalog No. 50-253-4473
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.