Learn More
Invitrogen™ Human Mesothelin ELISA Kit
Description
Human Mesothelin quantitates human Mesothelin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Mesothelin.
Specifications
Specifications
| Accession Number | CAK1; CAK1 antigen; megakaryocyte potentiating factor; Megakaryocyte-potentiating factor; Mes; Mesothelin; Mesothelin, cleaved form; MPF; Msln; pre-pro-megakaryocyte-potentiating factor; SMR; SMRP; soluble MPF mesothelin related protein |
| Assay Range | 100 to 30 ng/mL |
| Assay Sensitivity | 100 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 10232 |
| Gene Symbol | MSLN |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.