Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Metabolic Luminex™ Performance Premixed Kit

Catalog No. FCSTM1911
Change view
Click to view available options
Quantity:
1 Kit
10 Plex
11 Plex
12 Plex
2 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCSTM1911 11 Plex
FCSTM19 1 Kit
FCSTM1906 6 Plex
FCSTM1907 7 Plex
FCSTM1910 10 Plex
FCSTM1903 3 Plex
FCSTM1908 8 Plex
FCSTM1909 9 Plex
FCSTM1902 2 Plex
FCSTM1912 12 Plex
FCSTM1904 4 Plex
FCSTM1905 5 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. FCSTM1911 Supplier R&D Systems™ Supplier No. FCSTM1911
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Quantity 11 Plex
Species Human
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.