Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human METAP2 ELISA Kit

Catalog No. EH323RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH323RB 96 Tests
EH323RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH323RB Supplier Invitrogen™ Supplier No. EH323RB
Add to Cart
Add to Cart

ELISA

Human METAP2 quantitates human METAP2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human METAP2.

The ADP-ribosylation factor (Arf) family are highly conserved members of the Ras superfamily of regulatory GTP-binding proteins. Arf proteins participate in routing of intracellular proteins to and within the Golgi complex. Cellular functions include maintenance of organelle integrity, coat protein assembly, as an activator of phospholipase D. The Arf family is divided functionally into the Arf and the Arf-like (Arl) proteins. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6) and members of each class share a common gene organization.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 4930584B20Rik; A930035J23Rik; AI047573; AL024412; Amp2; AU014659; eIF-2-associated p67; eIF-2-associated p67 homolog; initiation factor 2 associated 67 kDa protein; initiation factor 2-associated 67 kDa glycoprotein; initiation factor 2-associated protein (p67); MAP 2; MAP2; metAP 2; METAP2; Methionine aminopeptidase 2; methionyl aminopeptidase 2; Mnpep; p67; p67eIF2; Peptidase M; peptidase M 2; similar to methionyl aminopeptidase 2; testicular tissue protein Li 17
Assay Range 2 to 250 ng/mL
Assay Sensitivity 1 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 10988
Gene Symbol METAP2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target METAP2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.