Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MFG-E8 ELISA Kit

Catalog No. EH324RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH324RBX10 10 x 96 Tests
EH324RBX5 5 x 96 Tests
EH324RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH324RBX10 Supplier Invitrogen™ Supplier No. EH324RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human MFG-E8 quantitates human MFG-E8 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human MFG-E8.

This gene encodes a preproprotein that is proteolytically processed to form multiple protein products. The major encoded protein product, lactadherin, is a membrane glycoprotein that promotes phagocytosis of apoptotic cells. This protein has also been implicated in wound healing, autoimmune disease, and cancer. Lactadherin can be further processed to form a smaller cleavage product, medin, which comprises the major protein component of aortic medial amyloid (AMA). Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AA408458; Ags; AI325141; BA46; Breast epithelial antigen BA46; EDIL1; EGF/factor VIII; HMFG; hP47; HsT19888; lactadherin; Lactadherin short form; Medin; Mfge8; MFG-E8; MFGM; MFGME8; MFGMP-E8; milk fat globule glycoprotein; milk fat globule membrane; milk fat globule membrane protein; Milk fat globule-EGF factor 8; milk fat globule-EGF factor 8 protein; MP47; O-acetyl disialoganglioside synthase; O-acetyl GD3 ganglioside synthase; O-acetyltransferase milk fat globule membrane protein; O-acetyltransferase Milk fat glolbule membrane protein; OAcGD3S; P47; SED1; SPAG10; sperm associated antigen 10; sperm surface protein hP47; Sperm surface protein SP47
Assay Range 0.1 to 25 ng/mL
Assay Sensitivity 0.1 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4240
Gene Symbol MFGE8
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target MFGE8 (Lactadherin)
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.