Learn More
R&D Systems™ Human mGluR8 (NP_000836) VersaClone cDNA
SDP
Catalog No. RDC1002 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
RDC1002 10 μg
1 options
This item is not returnable. View return policy
Catalog No. RDC1002 Supplier R&D Systems™ Supplier No. RDC1002
Add to Cart
Add to Cart
This item is not returnable. View return policy
Glutamate receptor, metabotropic 8
The Human mGluR8 (NP_000836) VersaClone cDNA clone contains the complete ORF for mGluR8, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The mGluR8 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans