Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human MIA-2 Recombinant Protein, PeproTech®

Catalog No. 1300220UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
130-02-20UG 20 μg
13002100UG 100 μg
130-02-1MG 1 mg
13002250UG 250 μg
13002500UG 500 μg
130-02-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 130-02-20UG Supplier Gibco™ Supplier No. 1300220UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human MIA-2 is a 101 amino acid, 11.5 kDa protein comprising the MIA homologous N-terminal region of the full length MIA-2 protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CTAGE5/MIA-2 is a secreted cytokine, and a member of the MIA/OTOR family. Members of this family, which also includes MIA, OTOR, and TANGO, share a Src homology-3 (SH3)-like domain. MIA-2 is predominantly expressed in hepatocytes. Elevated levels of MIA-2 may represent a clinically useful marker for diagnosis of hepatic disease activity and severity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96PC5
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 4253
Molecular Weight (g/mol) 11.5 kDa
Name Human MIA-2
Quantity 20 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias cTAGE family member 5; CTAGE family member 5 ER export factor; CTAGE family member 5, ER export factor; CTAGE family, member 5; CTAGE5; cutaneous T-cell lymphoma-associated antigen 5; LOW QUALITY PROTEIN: melanoma inhibitory activity protein 2; MEA11; MEA6; melanoma inhibitory activity 2; melanoma inhibitory activity protein 2; meningioma expressed antigen 6 (coiled-coil proline-rich); meningioma-expressed antigen 6/11; MGEA; MGEA11; MGEA6; MIA SH3 domain ER export factor 2; MIA2; protein cTAGE-5
Common Name CTAGE5
Gene Symbol Mia2
Conjugate Unconjugated
Recombinant Recombinant
Sequence MLESTKLLAD LKKCGDLECE ALINRVSAMR DYRGPDCRYL NFTKGEEISV YVKLAGERED LWAGSKGKEF GYFPRDAVQI EEVFISEEIQ MSTKESDFLC L
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.