Learn More
ACROBiosystems Human MICA 1mg
Supplier:  ACROBiosystems MIAH52551MG
Encompass_Preferred
Human MICA (24-202) Protein Mouse IgG2a Fc Tag (MIA-H5255) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Arg 202 (Accession # Q29983-1).
Catalog No. 50-253-4443
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.