Learn More
Biomatik Corporation Human Microalbunminuria (Mau/Alb) Elisa Kit
Supplier:  Biomatik Corporation EKC31286
Encompass_Preferred
Human Microalbunminuria (Mau/Alb) Elisa Kit for detecting Microalbunminuria(Mau/Alb) in serum, plasma, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 0.078-5ug/ml. Sensitivity: 0.019ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P02768; ALB. Target Synonyms: alb; ALBU_HUMAN; Albumin (32 AA); Albumin (AA 34); cell growth inhibiting protein 42; DKFZp779N1935; GIG20; GIG42; Growth inhibiting protein 20; growth-inhibiting protein 20; OTTHUMP00000220436; OTTHUMP00000220438; OTTHUMP00000220439; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7378
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.