Learn More
American Research Products Inc Human Microtubule associated p
Supplier:  American Research Products Inc CSBEL013403HU
Encompass
Human Microtubule-associated proteins 1A/1B light chain 3B (MAP1LC3B) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 67 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26994
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.