Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Midkine (MK) Recombinant Protein, PeproTech®

Catalog No. 4501650UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-16-50UG 50 μg
45016100UG 100 μg
450-16-1MG 1 mg
450-16-20UG 20 μg
45016250UG 250 μg
45016500UG 500 μg
450-16-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 450-16-50UG Supplier Gibco™ Supplier No. 4501650UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Midkine is a 13.4 kDa protein containing 123 amino acid residues including five intra-molecular disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21741
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 4192
Molecular Weight (g/mol) 13.4 kDa
Name Human Midkine (MK)
Quantity 50 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias amphiregulin-associated protein; ARAP; FLJ27379; MDK; Mek; Midgestation and kidney protein; midkine; midkine (neurite growth-promoting factor 2); MK; MK1; NEGF2; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein; Retanoic acid-responsive protein; retinoic acid inducible factor; retinoic acid-induced differentiation factor; retinoic acid-responsive protein
Common Name Midkine
Gene Symbol Mdk
Biological Activity Determined by its ability to chemoattract human neutrophils using a concentration range of 0.1-10.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence VAKKKDKVKK GGPGSECAEW AWGPCTPSSK DCGVGFREGT CGAQTQRIRC RVPCNWKKEF GADCKYKFEN WGACDGGTGT KVRQGTLKKA RYNAQCQETI RVTKPCTPKT KAKAKAKKGK GKD
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.