Learn More
FUJIFILM IRVINE SCIENTIFIC INC Human MIF, Recombinant Protein, Shenandoah Biotechnology, 25 ug vial
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 10015325UG
Encompass_Preferred
Macrophage migration inhibitory factor (MIF) is a pro-inflammatory lymphokine that functions during cell-mediated immmunity.  MIF promotes fibroblast migration by inducing interleukin 1 (IL-1), interleukin 8 (IL-8), and matrix metalloproteinase (MMP) expression.  In interferon-gamma-activated macrophages, MIF stimulates nitric oxide (NO) production and tumor necrosis factor-alpha (TNF-A) secretion. 
Catalog No. 50-900-514
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.