Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human MIF Recombinant Protein, PeproTech®

Catalog No. 300691MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-69-1MG 1 mg
30069100UG 100 μg
30069250UG 250 μg
300-69-25UG 25 μg
30069500UG 500 μg
300-69-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 300-69-1MG Supplier Gibco™ Supplier No. 300691MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Insect cell-derived Recombinant Human MIF is a 15 kDa protein containing 124 amino acid residues, including an N-terminal His-tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

MIF (macrophage migration inhibitory factor) was one of the first cytokine activities to be discovered and was initially described as a T cell-derived factor that inhibits the random migration of macrophages (Weiser 1989). Recently, MIF was rediscovered as a pituitary hormone that act as the counterregulatory hormone for glucocorticoid action within the immune system (Bernhagen 1993, Mitchell 1995). MIF was released from macrophages and T cells in response to physiological concentrations of glucocorticoids. The secreted MIF counter-regulates the immunosuppressive effects of steroids on immune cell activation and cytokine production (Bucala 1998). MIF plays a critical role in the host control of inflammation and immunity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14174
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 4282
Molecular Weight (g/mol) 15 kDa
Name Human MIF
Quantity 1 mg
Source Insect cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias cytokine; delayed early response protein 6; DER6; GIF; GLIF; glutathione-binding 13 kDa protein; Glycosylation-inhibiting factor; HGNC:7097; L-dopachrome isomerase; L-dopachrome tautomerase; macrophage migration inhibitory factor; macrophage migration inhibitory factor (glycosylation-inhibiting factor); MGC107654; Mif; MMIF; phenylpyruvate tautomerase
Common Name MIF
Gene Symbol Mif
Biological Activity Determined by its ability to inhibit monocyte migration.
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence HHHHHHHHAM PMFIVNTNVP RASVPDGFLS ELTQQLAQAT GKPPQYIAVH VVPDQLMAFG GSSEPCALCS LHSIGKIGGA QNRSYSKLLC GLLAERLRIS PDRVYINYYD MNAANVGWNN STFA
Content And Storage -20°C
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.