Learn More
FUJIFILM IRVINE SCIENTIFIC INC Human MIP-1b (CCL4), Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 100134100UG
Encompass_Preferred
Macrophage inflammatory protein-1 beta (MIP-1 B), also known as CCL4, is  produced by macrophages and functions as a mitogen-inducible cytokine.  MIP-1 B signals through the chemokine receptor CCR5 to chemoattract immune cells.  MIP-1 B induces inflammatory responses, including neutrophil superoxide production.  The MIP-1 B and MIP-1 A heterodimer exhibits antiviral activity against the human immunodeficiency virus 1 (HIV-1). 
Catalog No. 50-900-491
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.