Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human MIP-3 (CCL23) Recombinant Protein, PeproTech®

Catalog No. 30029250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30029250UG 250 μg
30029100UG 100 μg
300-29-1MG 1 mg
300-29-20UG 20 μg
30029500UG 500 μg
300-29-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 30029250UG Supplier Gibco™ Supplier No. 30029250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human MIP-3 is an 11.3 kDa protein containing 99 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. This chemokine, a member of the CC subfamily, displays chemotactic activity on resting T lymphocytes and monocytes, lower activity on neutrophils and no activity on activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. In addition, the product of this gene is a potent agonist of the chemokine (C-C motif) receptor 1. Alternative splicing results in multiple transcript variants that encode different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P55773
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6368
Molecular Weight (g/mol) 11.3 kDa
Name Human MIP-3 (CCL23)
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C6 beta-chemokine; C-C motif chemokine 23; C-C motif chemokine ligand 23; CCL23; CCL23(19-99); CCL23(22-99); CCL23(27-99); CCL23(30-99); chemokine (C-C motif) ligand 23; Ck beta 8-1; CKb8; CKB-8; Ckb-8-1; CK-BETA-8; hmrp-2 A; Macrophage inflammatory protein 3; MIP3; MIP-3; MIP-3 CK-BETA-8; MPIF1; MPIF-1; MPIF-1 Ckb-8-1; Myeloid progenitor inhibitory factor 1; SCYA23; small inducible cytokine subfamily A (Cys-Cys), member 23; Small-inducible cytokine A23
Common Name CCL23
Gene Symbol CCL23
Biological Activity Determined by its ability to chemoattract human T cell population using a concentration of 10-50 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence RVTKDAETEF MMSKLPLENP VLLDRFHATS ADCCISYTPR SIPCSLLESY FETNSECSKP GVIFLTKKGR RFCANPSDKQ VQVCMRMLKL DTRIKTRKN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.