Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Human MLKL Phospho S358 Synthetic Peptide, 1 mg
Description
- Target Protein: MLKL
- Type: Synthetic blocking peptide
- Tested Applications: BL
- Species Reactivity: Human
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | MLKL (phospho S358) |
| Substrate Properties | Peptide-based Substrate |
| Shipping Condition | Blue Ice |
| Label or Dye | Unconjugated |
| Reactivity | Human |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Form | Lyophilized |
| Cross Reactivity | Human |
| Quantity | 1 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?