Learn More
Gibco™ Human MMP-1 Recombinant Protein, PeproTech®
Description
420-01-1MG will be provided as 4 x 250 μg (420-01-250UG) 420-01-500UG will be provided as 2 x 250 μg (420-01-250UG) Recombinant Human MMP-1 is a 42.7 kDa protein containing the entire catalytic N-terminal domain and the C-terminal domain, which is involved in substrate specificity, and in binding TIMP-1. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P03956 |
| For Use With (Application) | Control, Enzyme Activity, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 4312 |
| Molecular Weight (g/mol) | 42.7 kDa |
| Name | Human MMP-1 |
| Purification Method | Purified |
| Quantity | 2 x 500 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.