Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human MMP-1 Recombinant Protein, PeproTech®

Catalog No. 4200150UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 250 μg
2 x 500 μg
250 μg
50 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
420-01-50UG 50 μg
42001100UG 100 μg
420-01-10UG 10 μg
42-001-01M 2 x 500 μg
42001250UG 250 μg
420-01-2UG 2 μg
42001500UG 2 x 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 420-01-50UG Supplier Gibco™ Supplier No. 4200150UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

420-01-1MG will be provided as 4 x 250 μg (420-01-250UG) 420-01-500UG will be provided as 2 x 250 μg (420-01-250UG) Recombinant Human MMP-1 is a 42.7 kDa protein containing the entire catalytic N-terminal domain and the C-terminal domain, which is involved in substrate specificity, and in binding TIMP-1. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P03956
For Use With (Application) Control, Enzyme Activity, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 4312
Molecular Weight (g/mol) 42.7 kDa
Name Human MMP-1
Purification Method Purified
Quantity 50 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 22 kDa interstitial collagenase; 27 kDa interstitial collagenase; CLG; CLGN; Collagenase; collagenase-like A; fibroblast collagenase; Interstitial collagenase; interstitial collagenase A; matrix metallo protease; matrix metallopeptidase 1; matrix metallopeptidase 1 (interstitial collagenase); matrix metallopeptidase 1a; matrix metallopeptidase 1a (interstitial collagenase); matrix metalloprotease 1; matrix metalloproteinase 1; matrix metalloproteinase 1a (interstitial collagenase); Matrix metalloproteinase1; Matrix metalloproteinase-1; Matrix metalloproteinase-1a; McolA; Mcol-A; MMP; MMP1; MMP-1; Mmp1a; MMP-1a; MMPs
Common Name MMP1
Gene Symbol MMP1
Biological Activity MMP-1 activity was measured by its ability to cleave a chromogenic peptide MMP-1 substrate at room temperature. At an MMP-1 concentration of 2.5 ug/ml, 50% cleavage was achieved (at an incubation time of approximately 25 minutes.)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MFVLTEGNPR WEQTHLTYRI ENYTPDLPRA DVDHAIEKAF QLWSNVTPLT FTKVSEGQAD IMISFVRGDH RDNSPFDGPG GNLAHAFQPG PGIGGDAHFD EDERWTNNFR EYNLHRVAAH ELGHSLGLSH STDIGALMYP SYTFSGDVQL AQDDIDGIQA IYGRSQNPVQ PIGPQTPKAC DSKLTFDAIT TIRGEVMFFK DRFYMRTNPF YPEVELNFIS VFWPQLPNGL EAAYEFADRD EVRFFKGNKY WAVQGQNVLH GYPKDIYSSF GFPRTVKHID AALSEENTGK TYFFVANKYW RYDEYKRSMD PGYPKMIAHD FPGIGHKVDA VFMKDGFFYF FHGTRQYKFD PKTKRILTLQ KANSWFNCRK N
Content And Storage -20°C
Activity MMP-1 activity was measured by its ability to cleave a chromogenic peptide MMP-1 substrate at room temperature. At an MMP-1 concentration of 2.5 ug/ml, 50% cleavage was achieved at an incubation time of approximately 25 minutes.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.