Learn More
Sino Biological Human MMP10 Protein 1MG 10249-H07H-1
Supplier:  Sino Biological 10249H07H1
Encompass_Preferred
A DNA sequence encoding the Human MMP10 (P09238) (Phe99-Cys476) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-241-1075
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.