Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MNAT1 Full-length ORF (NP_002422.1, 1 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89952716 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-952-716 25 ug
89-952-715 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-952-716 Supplier Abnova™ Supplier No. H00004331P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Cyclin-dependent kinases (CDKs), which play an essential role in cell cycle control of eukaryotic cells, are phosphorylated and thus activated by the CDK-activating kinase (CAK). CAK is a multisubunit protein that includes CDK7 (MIM 601955), cyclin H (CCNH; MIM 601953), and MAT1. MAT1 (for 'menage a trois-1') is involved in the assembly of the CAK complex.[supplied by OMIM]

Sequence: MDDQGCPRCKTTKYRNPSLKLMVNVCGHTLCESCVDLLFVRGAGNCPECGTPLRKSNFRVQLFEDPTVDKEVEIRKKVLKIYNKREEDFPSLREYNDFLEEVEEIVFNLTNNVDLDNTKKKMEIYQKENKDVIQKNKLKLTREQEELEEALEVERQENEQRRLFIQKEEQLQQILKRKNKQAFLDELESSDLPVALLLAQHKDRSTQLEMQLEKPKPVKPVTFSTGIKMGQHISLAPIHKLEEALYEYQPLQIETYGPHVPELEMLGRLGYLNHVRAASPQDLAGGYTSSLACHRALQDAFSGLFWQPS

Specifications

Accession Number NP_002422.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4331
Molecular Weight (g/mol) 62.2kDa
Name MNAT1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MDDQGCPRCKTTKYRNPSLKLMVNVCGHTLCESCVDLLFVRGAGNCPECGTPLRKSNFRVQLFEDPTVDKEVEIRKKVLKIYNKREEDFPSLREYNDFLEEVEEIVFNLTNNVDLDNTKKKMEIYQKENKDVIQKNKLKLTREQEELEEALEVERQENEQRRLFIQKEEQLQQILKRKNKQAFLDELESSDLPVALLLAQHKDRSTQLEMQLEKPKPVKPVTFSTGIKMGQHISLAPIHKLEEALYEYQPLQIETYGPHVPELEMLGRLGYLNHVRAASPQDLAGGYTSSLACHRALQDAFSGLFWQPS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MAT1/RNF66/TFB3
Common Name MNAT1
Gene Symbol MNAT1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.