Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human MOG ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP349837 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP349837 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP349837 Supplier Novus Biologicals™ Supplier No. NBP349837
Add to Cart
Add to Cart

Colorimetric Assay Kit

Specifications

Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Detection Method Chemiluminescence
Sample Type Serum, Plasma, Other Biological Fluids
Specificity This kit recognizes Human MOG in samples. No significant cross-reactivity or interference between Human MOG and analogues was observed
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 4340
Gene Symbol MOG
Quantity 1 Kit
Research Discipline Cellular Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.