Learn More
Kingfisher Biotech Inc Multi-species FGF basic (Human, Monkey) Recombinant Protein
Supplier: Kingfisher Biotech Inc RP1324HY005
The Human & Monkey FGF basic yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Human & Monkey FGF basic applications are for cell culture, ELISA standard, and Western Blot Control. Human & Monkey FGF basic yeast-derived recombinant protein can be purchased in multiple sizes. Human & Monkey FGF basic Specifications: (Molecular Weight: 16.3 kDa) (Amino Acid Sequence: ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS (145)) (Gene ID: n/a). For research use only. Made in the USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.