Learn More
Kingfisher Biotech Inc Human/Monkey VEGF-A (121AA) Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1610H500
Encompass_Preferred
The Human & Monkey VEGF-A yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Human & Monkey VEGF-A applications are for cell culture, ELISA standard, and Western Blot Control. Human & Monkey VEGF-A yeast-derived recombinant protein can be purchased in multiple sizes. Human & Monkey VEGF-A Specifications: (Molecular Weight: 14.1 kDa) (Amino Acid Sequence: APMAEGGGQN HHEVVKFMDV YQRSYCHPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCGGC CNDEGLECVP TEESNITMQI MRIKPHQGQH IGEMSFLQHN KCECRPKKDR ARQEKCDKPR R (121)) (Gene ID: 7422 (Human)). For research use only. Made in the USA
Catalog No. 50-253-3072
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.