Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Active Caspase-3 Antibody, R&D Systems™
GSA_VAGREENER_CHOICESDP

Catalog No. AF835
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF835 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF835 Supplier R&D Systems Supplier No. AF835

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 155 publications

Caspase-3 Polyclonal specifically detects Caspase-3 in Canine∣Chicken∣Cyprinus carpio (Carp)∣Equine∣Ewes∣Feline∣Fish∣Guinea Pig∣Human∣Mouse∣Oncorhynchus mykiss (Rainbow Trout)∣Ovine∣Pig∣Primate∣Rat∣Sheep∣Transgenic Mouse∣Xenograft∣Zebrafish samples. It is validated for ChIP Assay∣Flow Cytometry∣Functional Assay∣Immunocytochemistry∣Immunocytochemistry/Immunofluorescence∣Immunofluorescence∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Immunoprecipitation∣Western Blot.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen Caspase-3
Applications ChIP Assay, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. P42574
Gene Alias Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama
Gene Symbols CASP3
Host Species Rabbit
Immunogen KLH-coupled human Caspase-3 synthetic peptide CRGTELDCGIETD Accession # P42574
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 836
Test Specificity Distinguishes apoptotic cells from non-apoptotic cells in immunohistochemistry. Detects the p17 subunit of Caspase-3, but does not detect the precursor form in Western blot. We recommend R&D Systems Affinity Purified Anti-Caspase-3 (Catalog # AF-605-NA) or Monoclonal Anti-Cleaved-Caspase-3 (Catalog # MAB835) for Western blot applications.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Canine, Chicken, Carp, Equine, Ewe, Feline, Fish, Guinea Pig, Human, Mouse, Rainbow Trout, Ovine, Pig, Primate, Rat, Sheep, Transgenic Mouse, Xenograft, Zebrafish
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.