Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Aldehyde Dehydrogenase 1-A1/ ALDH1A1 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF5869SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5869SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5869SP Supplier R&D Systems Supplier No. AF5869SP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

Aldehyde Dehydrogenase 1-A1/ALDH1A1 Polyclonal specifically detects Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Human∣Mouse∣Transgenic Mouse∣Xenograft samples. It is validated for Flow Cytometry∣Immunocytochemistry∣Immunohistochemistry∣Immunoprecipitation∣Simple Western∣Western Blot.

Specifications

Antigen Aldehyde Dehydrogenase 1-A1/ALDH1A1
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Simple Western, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P00352
Gene Alias acetaldehyde dehydrogenase 1, ALDC, aldehyde dehydrogenase 1 family, member A1, aldehyde dehydrogenase 1, soluble, Aldehyde Dehydrogenase 1A1, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic, aldehyde dehydrogenase, liver cytosolic, ALDH class 1, ALDH1, ALDH11, ALDH1A1, ALDH-E1, ALHDII, EC 1.2.1, EC 1.2.1.36, MGC2318, PUMB1, RALDH 1, RalDH1, retinal dehydrogenase 1, retinaldehyde dehydrogenase 1
Gene Symbols ALDH1A1
Host Species Goat
Immunogen E. coli-derived recombinant human Aldehyde Dehydrogenase 1-A1/ALDH1A1 Ser2-Ser501 Accession # P00352
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216
Test Specificity Detects human Aldehyde Dehydrogenase 1-A1/ALDH1A1 in direct ELISAs and detects human and mouse Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Transgenic Mouse, Xenograft
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.