Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse c-Myc Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3696
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3696 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3696 Supplier R&D Systems Supplier No. AF3696
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

c-Myc Polyclonal specifically detects c-Myc in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, Knockout Validated.

Specifications

Antigen c-Myc
Applications ChIP Assay, Immunocytochemistry, Immunohistochemistry, KnockDown
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-15 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL, Knockout Validated
Gene Accession No. P01106
Gene Alias avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, cMyc, c-Myc, MYC, myc proto-oncogene protein, Myc2, MYCC, myc-related translation/localization regulatory factor, Niard, Nird, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)
Gene Symbols MYC
Host Species Goat
Immunogen E. coli-derived recombinant human c-Myc Arg66-Asp201 Accession # P01106
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4609
Test Specificity Detects human c-Myc in direct ELISAs. Detects human and mouse c-Myc in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) L-Myc and rhN-Myc is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.